Product Details

SNP ID
rs4877724
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:82981122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAATAAATGTTTTTAGCTTATA[C/T]AATACAATAAATAACAGCTACTTTT
Phenotype
MIM: 611344
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RASEF PubMed Links
Additional Information
For this assay, SNP(s) [rs116715552] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RASEF
Gene Name
RAS and EF-hand domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152573.3 4019 UTR 3 NP_689786.2
XM_005251730.3 4019 UTR 3 XP_005251787.1
XM_005251731.3 4019 UTR 3 XP_005251788.1

View Full Product Details