Product Details
- SNP ID
-
rs4513029
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:72970126 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCGGGACAGGAAGCCCTGCTGGA[A/G]AAGGCTCAGTTCCTTCCTTGCTGTT
- Phenotype
-
MIM: 616372
MIM: 605471
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DCAF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76102681] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCAF4
- Gene Name
- DDB1 and CUL4 associated factor 4
There are no transcripts associated with this gene.
- Gene
- ZFYVE1
- Gene Name
- zinc finger FYVE-type containing 1
View Full Product Details