Product Details

SNP ID
rs4402688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23652891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAAAGGCTTATATTTTCTTAAA[A/G]TCTTTAGACAGTAATTGCATTTATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 2842 Intron NP_612203.2
XM_017026442.1 2842 UTR 3 XP_016881931.1
XM_017026443.1 2842 UTR 3 XP_016881932.1
XM_017026444.1 2842 Intron XP_016881933.1

View Full Product Details