Product Details

SNP ID
rs4970632
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:38838323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTAAATACTCAAGTCAGAATCA[C/T]CTGCAGGAAGCACTCCAAGTCACAC
Phenotype
MIM: 608267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRAGC PubMed Links

Gene Details

Gene
RRAGC
Gene Name
Ras related GTP binding C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271851.1 2505 UTR 3 NP_001258780.1
NM_022157.3 2505 UTR 3 NP_071440.1

View Full Product Details