Product Details
- SNP ID
-
hCV28012639
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46671175 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCACCTCCCAACCCAGAGGAGG[C/G]GTGTCTCCAACTCAGCCCTCAGCCA
- Phenotype
-
MIM: 612402
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ALS2CL
PubMed Links
Gene Details
- Gene
- ALS2CL
- Gene Name
- ALS2 C-terminal like
View Full Product Details