Product Details

SNP ID
rs4821767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38218122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTACAGCCAAGTGGAAACTATCTT[A/C]CCCATGTTTCGGAATGCCCAGGGAG
Phenotype
MIM: 604877
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAFF PubMed Links

Gene Details

Gene
MAFF
Gene Name
MAF bZIP transcription factor F
There are no transcripts associated with this gene.

Gene
SNORA92
Gene Name
small nucleolar RNA, H/ACA box 92
There are no transcripts associated with this gene.

Gene
TMEM184B
Gene Name
transmembrane protein 184B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195071.1 Intron NP_001182000.1
NM_001195072.1 Intron NP_001182001.1
NM_012264.4 Intron NP_036396.2
XM_011530112.2 Intron XP_011528414.2
XM_011530113.1 Intron XP_011528415.1
XM_011530114.1 Intron XP_011528416.1
XM_011530115.2 Intron XP_011528417.1
XM_011530116.2 Intron XP_011528418.2
XM_011530117.1 Intron XP_011528419.1
XM_011530118.1 Intron XP_011528420.1
XM_011530119.1 Intron XP_011528421.1
XM_017028754.1 Intron XP_016884243.1
XM_017028755.1 Intron XP_016884244.1
XM_017028756.1 Intron XP_016884245.1
XM_017028757.1 Intron XP_016884246.1
XM_017028758.1 Intron XP_016884247.1
XM_017028759.1 Intron XP_016884248.1

View Full Product Details