Product Details

SNP ID
rs4073177
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:1069600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCAGGGCTGAGCCGCTGCTCC[G/A]CGCGCAGAACCCCGCAGCCCTGGGA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC100288175 PubMed Links

Gene Details

Gene
LOC100288175
Gene Name
uncharacterized LOC100288175
There are no transcripts associated with this gene.

Gene
LOC105378948
Gene Name
uncharacterized LOC105378948
There are no transcripts associated with this gene.

Gene
RNF223
Gene Name
ring finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205252.1 3268 Intron NP_001192181.1
XM_017001286.1 3268 UTR 3 XP_016856775.1
XM_017001287.1 3268 Intron XP_016856776.1

View Full Product Details