Product Details
- SNP ID
-
rs7249699
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23740234 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTAAAACCAATCTTGCAGTGCATT[A/T]AAAATACCGATTTCTCATCAAAACC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF681
PubMed Links
Gene Details
- Gene
- ZNF681
- Gene Name
- zinc finger protein 681
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138286.2 |
5458 |
UTR 3 |
|
|
NP_612143.2 |
View Full Product Details