Product Details

SNP ID
rs7821559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:9555790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCCGTTCCCTTGGCTGTTCTCTG[A/G]GCTCCCAATACTTCCCTCCTTCTCT
Phenotype
MIM: 603303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNKS PubMed Links

Gene Details

Gene
TNKS
Gene Name
tankyrase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003747.2 562 Intron NP_003738.2
XM_006716263.3 562 UTR 5 XP_006716326.1
XM_011543845.2 562 UTR 5 XP_011542147.1
XM_011543846.2 562 UTR 5 XP_011542148.1
XM_011543847.2 562 UTR 5 XP_011542149.1

View Full Product Details