Product Details

SNP ID
rs7561616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99290769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACTAAGAATTGTATAGAAAAAG[A/G]TAAAGGAAAATAAATTTAACATTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYG1 PubMed Links

Gene Details

Gene
LYG1
Gene Name
lysozyme g1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174898.2 Intron NP_777558.1
XM_005263869.2 Intron XP_005263926.1
XM_005263870.3 Intron XP_005263927.1
XM_005263871.4 Intron XP_005263928.1
XM_011510580.2 Intron XP_011508882.1
XM_017003311.1 Intron XP_016858800.1
XM_017003312.1 Intron XP_016858801.1
XM_017003313.1 Intron XP_016858802.1

View Full Product Details