Product Details

SNP ID
rs4835010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145632151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTCACTAATGTCCTTTTCTTTCT[C/T]GCTGAAGTGTCCTCTTTAGCATTTC
Phenotype
MIM: 607481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMAA PubMed Links

Gene Details

Gene
MMAA
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172250.2 Intron NP_758454.1
XM_011531684.2 Intron XP_011529986.1
XM_011531685.2 Intron XP_011529987.1
XM_011531686.2 Intron XP_011529988.1

View Full Product Details