Product Details

SNP ID
rs7904886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49301193 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACTCCATATGCCCTGGGTTACT[A/G]GAGGGAAGCAAAGGGCTTTGAGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf71 PubMed Links

Gene Details

Gene
C10orf71
Gene Name
chromosome 10 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135196.1 Intron NP_001128668.1
XM_005269476.4 Intron XP_005269533.1
XM_005269477.3 Intron XP_005269534.1
XM_005269478.4 Intron XP_005269535.1
XM_017015619.1 Intron XP_016871108.1
Gene
C10orf71-AS1
Gene Name
C10orf71 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details