Product Details

SNP ID
rs8074245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60424900 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAATTCCATAGACCCTGTTACAAG[A/G]ATACTCCCTTAACTGCTCTGCAGAT
Phenotype
MIM: 607740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf64 PubMed Links

Gene Details

Gene
C17orf64
Gene Name
chromosome 17 open reading frame 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181707.2 Intron NP_859058.2
XM_005257033.4 Intron XP_005257090.1
XM_005257034.4 Intron XP_005257091.1
XM_005257035.4 Intron XP_005257092.1
XM_005257036.2 Intron XP_005257093.1
XM_017024166.1 Intron XP_016879655.1
Gene
RPL12P38
Gene Name
ribosomal protein L12 pseudogene 38
There are no transcripts associated with this gene.

Gene
USP32
Gene Name
ubiquitin specific peptidase 32
There are no transcripts associated with this gene.

View Full Product Details