Product Details

SNP ID
rs6671190
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:114579346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACCTGGGAGTTTACTAATTAAAT[A/C]ATCAGTAAACTAATTACAAAGGTAT
Phenotype
MIM: 605783
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCAS2 PubMed Links

Gene Details

Gene
BCAS2
Gene Name
breast carcinoma amplified sequence 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005872.2 Intron NP_005863.1
Gene
DENND2C
Gene Name
DENN domain containing 2C
There are no transcripts associated with this gene.

View Full Product Details