Product Details

SNP ID
rs6802114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150604453 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATGACATTTTCTCAGGATAAAC[C/T]GACCATCCCAGAATAGCCAGATTAG
Phenotype
MIM: 607912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SELT PubMed Links

Gene Details

Gene
SELT
Gene Name
selenoprotein T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016275.3 Intron NP_057359.2
Gene
SERP1
Gene Name
stress-associated endoplasmic reticulum protein 1
There are no transcripts associated with this gene.

View Full Product Details