Product Details

SNP ID
rs6790721
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128913611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGCAGCCGACTCTTTTTCCTCC[A/G]AGTATATTCCCAATTTACTGTGATG
Phenotype
MIM: 611103
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACAD9 PubMed Links

Gene Details

Gene
ACAD9
Gene Name
acyl-CoA dehydrogenase family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014049.4 5164 Intron NP_054768.2
XM_017006264.1 5164 Intron XP_016861753.1
XM_017006265.1 5164 Intron XP_016861754.1
Gene
KIAA1257
Gene Name
KIAA1257
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020741.2 5164 Intron NP_065792.1
XM_011513050.2 5164 Intron XP_011511352.2
XM_017006939.1 5164 UTR 3 XP_016862428.1
XM_017006940.1 5164 UTR 3 XP_016862429.1
XM_017006941.1 5164 UTR 3 XP_016862430.1
XM_017006942.1 5164 UTR 3 XP_016862431.1
XM_017006943.1 5164 UTR 3 XP_016862432.1
XM_017006944.1 5164 UTR 3 XP_016862433.1
XM_017006945.1 5164 Intron XP_016862434.1
XM_017006946.1 5164 UTR 3 XP_016862435.1
XM_017006947.1 5164 UTR 3 XP_016862436.1
XM_017006948.1 5164 UTR 3 XP_016862437.1
XM_017006949.1 5164 UTR 3 XP_016862438.1
XM_017006950.1 5164 UTR 3 XP_016862439.1
XM_017006951.1 5164 UTR 3 XP_016862440.1
XM_017006952.1 5164 UTR 3 XP_016862441.1
XM_017006953.1 5164 UTR 3 XP_016862442.1
XM_017006954.1 5164 Intron XP_016862443.1

View Full Product Details