Product Details

SNP ID
rs7780748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116878310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCATAGAAGACTTACGCTGTATT[A/G]GGAGGCAGCTATGTAAAAAGCTGAG
Phenotype
MIM: 601571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPZA2 PubMed Links

Gene Details

Gene
CAPZA2
Gene Name
capping actin protein of muscle Z-line alpha subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006136.2 Intron NP_006127.1

View Full Product Details