Product Details

SNP ID
rs7760132
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55434753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATGTGGGTAAGCAAAGCACCA[C/T]GAATAAAAGGAAAATGGGTATTTCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HMGCLL1 PubMed Links

Gene Details

Gene
HMGCLL1
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042406.1 1995 UTR 3 NP_001035865.1
NM_001287741.1 1995 UTR 3 NP_001274670.1
NM_001287746.1 1995 UTR 3 NP_001274675.1
NM_001287753.1 1995 UTR 3 NP_001274682.1
NM_019036.2 1995 UTR 3 NP_061909.2
XM_011514700.2 1995 Intron XP_011513002.1

View Full Product Details