Product Details

SNP ID
rs7982436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:98795176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACGTGAGGTCTACTTAAGGCAAC[G/T]TGAGGTCTACTTACATGTGCTTGAC
Phenotype
MIM: 607325
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DOCK9 PubMed Links

Gene Details

Gene
DOCK9
Gene Name
dedicator of cytokinesis 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130048.1 Intron NP_001123520.1
NM_001130049.1 Intron NP_001123521.1
NM_001130050.1 Intron NP_001123522.1
NM_001318849.1 Intron NP_001305778.1
NM_015296.2 Intron NP_056111.1
XM_005254034.3 Intron XP_005254091.1
XM_005254035.3 Intron XP_005254092.1
XM_006719922.1 Intron XP_006719985.1
XM_006719924.2 Intron XP_006719987.1
XM_006719925.1 Intron XP_006719988.1
XM_006719931.3 Intron XP_006719994.1
XM_006719932.1 Intron XP_006719995.1
XM_006719936.1 Intron XP_006719999.1
XM_006719937.3 Intron XP_006720000.1
XM_017020503.1 Intron XP_016875992.1
XM_017020504.1 Intron XP_016875993.1
XM_017020505.1 Intron XP_016875994.1
XM_017020506.1 Intron XP_016875995.1
XM_017020507.1 Intron XP_016875996.1
XM_017020508.1 Intron XP_016875997.1
XM_017020509.1 Intron XP_016875998.1
XM_017020510.1 Intron XP_016875999.1
XM_017020511.1 Intron XP_016876000.1
XM_017020512.1 Intron XP_016876001.1
XM_017020513.1 Intron XP_016876002.1
XM_017020514.1 Intron XP_016876003.1
XM_017020515.1 Intron XP_016876004.1
XM_017020516.1 Intron XP_016876005.1
XM_017020517.1 Intron XP_016876006.1
XM_017020518.1 Intron XP_016876007.1
XM_017020519.1 Intron XP_016876008.1
XM_017020520.1 Intron XP_016876009.1
XM_017020521.1 Intron XP_016876010.1

View Full Product Details