Product Details

SNP ID
rs7026378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127909827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAGCGCAAGGGGTATCTGTGCT[A/G]TGGTCACCCACCATGAAGCCCACAC
Phenotype
MIM: 606378 MIM: 610135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST6GALNAC4 PubMed Links

Gene Details

Gene
ST6GALNAC4
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175039.3 Intron NP_778204.1
NM_175040.3 Intron NP_778205.1
XM_011518527.2 Intron XP_011516829.1
XM_011518529.2 Intron XP_011516831.1
Gene
ST6GALNAC6
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details