Product Details
- SNP ID
-
rs8112910
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:36608658 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTCTAGTGACTCTCAAATACCC[C/G]TCTCCACTCTAGACCTCTGTCCAGA
- Phenotype
-
MIM: 609516
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF382
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149550922,rs78618245] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF382
- Gene Name
- zinc finger protein 382
- Gene
- ZNF529
- Gene Name
- zinc finger protein 529
There are no transcripts associated with this gene.
View Full Product Details