Product Details

SNP ID
rs7301339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56204890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCATTACATGGTGGGGAGGAGCC[C/T]TTCTATCTCCAACCACACTCCCACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF41 PubMed Links

Gene Details

Gene
RNF41
Gene Name
ring finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242826.1 2942 UTR 3 NP_001229755.1
NM_005785.3 2942 UTR 3 NP_005776.1
NM_194358.2 2942 UTR 3 NP_919339.1
NM_194359.2 2942 UTR 3 NP_919340.1
XM_005268561.4 2942 UTR 3 XP_005268618.1
XM_011537734.2 2942 UTR 3 XP_011536036.1
XM_011537735.2 2942 UTR 3 XP_011536037.1

View Full Product Details