Product Details

SNP ID
rs7461531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:15545181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCATCTATCTTGGTGGTCAGT[A/T]TTACTGTCATAACTATTAAATATTT
Phenotype
MIM: 601385
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TUSC3 PubMed Links

Gene Details

Gene
TUSC3
Gene Name
tumor suppressor candidate 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006765.3 Intron NP_006756.2
NM_178234.2 Intron NP_839952.1
XM_011544651.2 Intron XP_011542953.1
XM_011544652.2 Intron XP_011542954.1
XM_011544654.2 Intron XP_011542956.1
XM_017013860.1 Intron XP_016869349.1
XM_017013861.1 Intron XP_016869350.1

View Full Product Details