Product Details

SNP ID
rs7900561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15097648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTCCTCTGCTGGATTGCAGCGC[C/T]GGCTGGGCACCAGGAGTACAAAGCG
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACBD7 PubMed Links

Gene Details

Gene
ACBD7
Gene Name
acyl-CoA binding domain containing 7
There are no transcripts associated with this gene.

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153244.1 234 Intron NP_694976.1
Gene
NMT2
Gene Name
N-myristoyltransferase 2
There are no transcripts associated with this gene.

Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097590.2 234 Intron NP_001091059.1
NM_001265601.1 234 UTR 5 NP_001252530.1
NM_006414.4 234 Intron NP_006405.2
NM_183005.4 234 UTR 5 NP_892117.1
XM_006717363.1 234 UTR 5 XP_006717426.1
XM_006717364.3 234 Intron XP_006717427.1
XM_011519293.1 234 UTR 5 XP_011517595.1
XM_017015479.1 234 UTR 5 XP_016870968.1
XM_017015480.1 234 Intron XP_016870969.1
XM_017015481.1 234 Intron XP_016870970.1

View Full Product Details