Product Details
- SNP ID
-
rs7353835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:5915428 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTATGGACTTGATGCTGTACTAGG[A/G]GGGTTAGATATCTGCCATGTTCTTT
- Phenotype
-
MIM: 118920
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHGB
PubMed Links
Gene Details
- Gene
- CHGB
- Gene Name
- chromogranin B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001819.2 |
|
Intron |
|
|
NP_001810.2 |
View Full Product Details