Product Details

SNP ID
rs6865399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135338889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTGCTTTGGGCCAGGGTGTTAA[C/G]CTCTCTAAGCATTAATTTCCTCCAC
Phenotype
MIM: 610054
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5orf66 PubMed Links

Gene Details

Gene
C5orf66
Gene Name
chromosome 5 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277348.1 Intron NP_001264277.1
Gene
H2AFY
Gene Name
H2A histone family member Y
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040158.1 Intron NP_001035248.1
NM_004893.2 Intron NP_004884.1
NM_138609.2 Intron NP_613075.1
NM_138610.2 Intron NP_613258.2
XM_005272132.1 Intron XP_005272189.1
XM_005272134.3 Intron XP_005272191.1
XM_005272135.3 Intron XP_005272192.1
XM_011543729.2 Intron XP_011542031.1
XM_011543730.2 Intron XP_011542032.1
XM_011543731.1 Intron XP_011542033.1
XM_011543732.1 Intron XP_011542034.1
XM_011543733.2 Intron XP_011542035.1
XM_011543734.2 Intron XP_011542036.1
XM_011543735.2 Intron XP_011542037.1
XM_017010067.1 Intron XP_016865556.1
XM_017010068.1 Intron XP_016865557.1
XM_017010069.1 Intron XP_016865558.1

View Full Product Details