Product Details

SNP ID
rs7987725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27549310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTATATAGAATGTCAACATTTA[C/T]TTTTTAAGAACTGGTTCTTCCTTCG
Phenotype
MIM: 609733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LNX2 PubMed Links

Gene Details

Gene
LNX2
Gene Name
ligand of numb-protein X 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153371.3 Intron NP_699202.1
XM_011534995.2 Intron XP_011533297.1
XM_017020434.1 Intron XP_016875923.1

View Full Product Details