Product Details

SNP ID
rs7756024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125958042 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGGTAGCCAGTTACAATGCCTC[G/T]TGCTGTGGTCAGGTGATTTTACAGT
Phenotype
MIM: 609998
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HINT3 PubMed Links

Gene Details

Gene
HINT3
Gene Name
histidine triad nucleotide binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138571.4 Intron NP_612638.3

View Full Product Details