Product Details

SNP ID
rs8128151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29033442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTCTCCAAAGTATATATTTAGC[G/T]GCAATGGAAACACGACCGGTTGTTG
Phenotype
MIM: 604735
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
USP16 PubMed Links

Gene Details

Gene
USP16
Gene Name
ubiquitin specific peptidase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001992.1 Intron NP_001001992.1
NM_001032410.1 Intron NP_001027582.1
NM_006447.2 Intron NP_006438.1
XM_017028257.1 Intron XP_016883746.1
XM_017028258.1 Intron XP_016883747.1
XM_017028259.1 Intron XP_016883748.1
XM_017028260.1 Intron XP_016883749.1
XM_017028261.1 Intron XP_016883750.1

View Full Product Details