Product Details

SNP ID
rs5974536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135530753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACCTGTCATAGCCAGATCATTGG[G/T]TTTTTCCCCTCCTGTGTAGTTAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
INTS6L PubMed Links

Gene Details

Gene
INTS6L
Gene Name
integrator complex subunit 6 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182540.4 Intron NP_872346.3
XM_005262394.2 Intron XP_005262451.1
XM_006724740.2 Intron XP_006724803.1
XM_006724741.3 Intron XP_006724804.1
XM_011531311.2 Intron XP_011529613.1
XM_017029342.1 Intron XP_016884831.1
XM_017029343.1 Intron XP_016884832.1
XM_017029344.1 Intron XP_016884833.1
XM_017029345.1 Intron XP_016884834.1
XM_017029346.1 Intron XP_016884835.1
XM_017029347.1 Intron XP_016884836.1
XM_017029348.1 Intron XP_016884837.1
XM_017029349.1 Intron XP_016884838.1

View Full Product Details