Product Details
- SNP ID
-
rs6141898
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:33238324 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCGACCCTGAAGCAGCGAGGGAGC[G/T]GCCTGAAGATGGAGCCAGGACTCAG
- Phenotype
-
MIM: 607412
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BPIFA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1321418] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BPIFA1
- Gene Name
- BPI fold containing family A member 1
View Full Product Details