Product Details
- SNP ID
-
rs7527503
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:63448441 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCACTTGGTGTGAATAGTATGAA[C/T]AAAAATGACTACCCTCCTCTTATCT
- Phenotype
-
MIM: 604566
MIM: 605494
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALG6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76225126] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALG6
- Gene Name
- ALG6, alpha-1,3-glucosyltransferase
There are no transcripts associated with this gene.
- Gene
- ITGB3BP
- Gene Name
- integrin subunit beta 3 binding protein
View Full Product Details