Product Details

SNP ID
rs6937834
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52792426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATACAGTGTGGGGGGCCAGTGT[C/T]CTGGAGAGCTGTGCAGAGAGGAACA
Phenotype
MIM: 138359
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GSTA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs112846024] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GSTA1
Gene Name
glutathione S-transferase alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319059.1 1048 Intron NP_001305988.1
NM_145740.4 1048 Intron NP_665683.1
XM_005249034.3 1048 UTR 3 XP_005249091.1

View Full Product Details