Product Details

SNP ID
rs7639294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150551316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGATTTTCTTTCCTATTATTTC[G/T]CCAATGTTCAACACAGGGCCTAGGA
Phenotype
MIM: 609234
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF2A PubMed Links

Gene Details

Gene
EIF2A
Gene Name
eukaryotic translation initiation factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319043.1 Intron NP_001305972.1
NM_001319044.1 Intron NP_001305973.1
NM_001319045.1 Intron NP_001305974.1
NM_001319046.1 Intron NP_001305975.1
NM_032025.4 Intron NP_114414.2
XM_011513224.1 Intron XP_011511526.1
Gene
SERP1
Gene Name
stress-associated endoplasmic reticulum protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014445.3 Intron NP_055260.1
XM_011512698.2 Intron XP_011511000.1

View Full Product Details