Product Details

SNP ID
rs10512786
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:130683704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGCTATTATTTATCAAGCATTA[A/G]TTGTGTGCTAGAGTATGTATTTTGC
Phenotype
MIM: 616613 MIM: 602610
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
COL6A6 PubMed Links
Additional Information
For this assay, SNP(s) [rs78693657] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
COL6A6
Gene Name
collagen type VI alpha 6
There are no transcripts associated with this gene.

Gene
PIK3R4
Gene Name
phosphoinositide-3-kinase regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014602.2 Intron NP_055417.1

View Full Product Details