Product Details

SNP ID
rs7525580
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:154987406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGGCTGGAAAAGAAGATAGGGT[A/G]TACGCCACAGACAGCCCCAAATACT
Phenotype
MIM: 116900 MIM: 610595 MIM: 607377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CKS1B PubMed Links

Gene Details

Gene
CKS1B
Gene Name
CDC28 protein kinase regulatory subunit 1B
There are no transcripts associated with this gene.

Gene
FLAD1
Gene Name
flavin adenine dinucleotide synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184891.1 Intron NP_001171820.1
NM_001184892.1 Intron NP_001171821.1
NM_025207.4 Intron NP_079483.3
NM_201398.2 Intron NP_958800.1
Gene
LENEP
Gene Name
lens epithelial protein
There are no transcripts associated with this gene.

View Full Product Details