Product Details

SNP ID
rs10419451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14053411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCAAGTATAAAGGCTTCATCG[C/T]AGAAGGAGGCCAGGGTTACAGGCAG
Phenotype
MIM: 605350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004843.3 2216 Intron NP_004834.1
Gene
PALM3
Gene Name
paralemmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145028.1 2216 UTR 3 NP_001138500.1
XM_005259891.4 2216 UTR 3 XP_005259948.1
XM_011527965.2 2216 UTR 3 XP_011526267.1

View Full Product Details