Product Details

SNP ID
rs9863229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:167480525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCCAGTGTAAACTATACTGTA[A/G]AGAGGAAGTTATATAGACCTTGGCT
Phenotype
MIM: 605587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINI2 PubMed Links

Gene Details

Gene
SERPINI2
Gene Name
serpin family I member 2
There are no transcripts associated with this gene.

Gene
WDR49
Gene Name
WD repeat domain 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178824.3 Intron NP_849146.1

View Full Product Details