Product Details

SNP ID
rs10143409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94584328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGAGCTTACTTTTGGTGAAGGT[C/T]AAACATAGTAATATTTATGGAGTGT
Phenotype
MIM: 601841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA5 PubMed Links

Gene Details

Gene
SERPINA5
Gene Name
serpin family A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000624.5 Intron NP_000615.3

View Full Product Details