Product Details
- SNP ID
-
rs9887794
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:202133884 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGCCAGGACAAGCCAGATTGGGC[A/G]GTGGAAACACACGGGACAGGGAGGC
- Phenotype
-
MIM: 616597
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARL8A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114950674] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARL8A
- Gene Name
- ADP ribosylation factor like GTPase 8A
- Gene
- GPR37L1
- Gene Name
- G protein-coupled receptor 37 like 1
View Full Product Details