Product Details

SNP ID
rs9833732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195747110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTTTCCAGTCTCCCAAAAGCA[A/G]TGGCGCCTTAAATGTGCGGTAAGGA
Phenotype
MIM: 158372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC4 PubMed Links

Gene Details

Gene
MUC4
Gene Name
mucin 4, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004532.5 3753 UTR 3 NP_004523.3
NM_018406.6 3753 Intron NP_060876.5
NM_138297.4 3753 UTR 3 NP_612154.2

View Full Product Details