Product Details
- SNP ID
-
rs10146011
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:88616518 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGTTTGGTGAAAAGCTAATTACA[A/G]CTTTTGTAGGATGGTTCCAAAGATG
- Phenotype
-
MIM: 613279
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EML5
PubMed Links
Gene Details
- Gene
- EML5
- Gene Name
- echinoderm microtubule associated protein like 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183387.2 |
5939 |
Intron |
|
|
NP_899243.1 |
XM_006720070.3 |
5939 |
Intron |
|
|
XP_006720133.1 |
XM_011536528.2 |
5939 |
Silent Mutation |
CTG,TTG |
L2002L |
XP_011534830.1 |
XM_011536530.2 |
5939 |
Silent Mutation |
CTG,TTG |
L1999L |
XP_011534832.1 |
XM_011536531.2 |
5939 |
Silent Mutation |
CTG,TTG |
L1994L |
XP_011534833.1 |
XM_011536532.2 |
5939 |
Silent Mutation |
CTG,TTG |
L1994L |
XP_011534834.1 |
XM_011536533.2 |
5939 |
Silent Mutation |
CTG,TTG |
L1956L |
XP_011534835.1 |
XM_011536534.2 |
5939 |
Silent Mutation |
CTG,TTG |
L1943L |
XP_011534836.1 |
XM_011536536.2 |
5939 |
Intron |
|
|
XP_011534838.1 |
XM_011536538.2 |
5939 |
Intron |
|
|
XP_011534840.1 |
XM_017021062.1 |
5939 |
Intron |
|
|
XP_016876551.1 |
XM_017021063.1 |
5939 |
Silent Mutation |
CTG,TTG |
L1995L |
XP_016876552.1 |
XM_017021064.1 |
5939 |
Silent Mutation |
CTG,TTG |
L1964L |
XP_016876553.1 |
XM_017021065.1 |
5939 |
Intron |
|
|
XP_016876554.1 |
XM_017021066.1 |
5939 |
Silent Mutation |
CTG,TTG |
L1946L |
XP_016876555.1 |
XM_017021067.1 |
5939 |
Silent Mutation |
CTG,TTG |
L1945L |
XP_016876556.1 |
XM_017021068.1 |
5939 |
Silent Mutation |
CTG,TTG |
L1944L |
XP_016876557.1 |
XM_017021069.1 |
5939 |
Silent Mutation |
CTG,TTG |
L1943L |
XP_016876558.1 |
XM_017021070.1 |
5939 |
Intron |
|
|
XP_016876559.1 |
- Gene
- ZC3H14
- Gene Name
- zinc finger CCCH-type containing 14
There are no transcripts associated with this gene.
View Full Product Details