Product Details

SNP ID
rs6107716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:663275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCATTGGGGGCTCCAGTAGAGGAA[C/T]GGTCAGAGAGATTCAGCTGAGAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCRT2 PubMed Links

Gene Details

Gene
SCRT2
Gene Name
scratch family transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033129.3 1898 UTR 3 NP_149120.1
Gene
SRXN1
Gene Name
sulfiredoxin 1
There are no transcripts associated with this gene.

View Full Product Details