Product Details
- SNP ID
-
rs6860934
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:96666138 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATAATTAAAAGACATGTCTCAGAT[A/G]TGTTCGTTTAACAGCATGCCACCTA
- Phenotype
-
MIM: 114090
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAST
PubMed Links
Gene Details
- Gene
- CAST
- Gene Name
- calpastatin
View Full Product Details