Product Details

SNP ID
rs5945108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154676120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCATGTGTGTTTCTTCATGCTAT[A/G]AATACGTTTCTGGAAATGTTTGGTA
Phenotype
MIM: 300482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAB3 PubMed Links

Gene Details

Gene
GAB3
Gene Name
GRB2 associated binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081573.2 3732 UTR 3 NP_001075042.1
NM_001282283.1 3732 UTR 3 NP_001269212.1
NM_080612.3 3732 UTR 3 NP_542179.1
XM_005274648.1 3732 Intron XP_005274705.1
XM_006724804.1 3732 UTR 3 XP_006724867.1
XM_011531103.1 3732 UTR 3 XP_011529405.1
XM_011531105.1 3732 Intron XP_011529407.1
XM_011531106.1 3732 UTR 3 XP_011529408.1
XM_017029276.1 3732 UTR 3 XP_016884765.1

View Full Product Details