Product Details
- SNP ID
-
rs6685048
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:27782144 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGAAAGGGTCTTGCTCTGTCACC[C/G]AGGATGTGCCAGTAGTACAGTGGTA
- Phenotype
-
MIM: 606892
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STX12
PubMed Links
Gene Details
- Gene
- STX12
- Gene Name
- syntaxin 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177424.2 |
|
Intron |
|
|
NP_803173.1 |
View Full Product Details