Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142397.1 | Intron | NP_001135869.1 | |||
NM_014772.2 | Intron | NP_055587.1 | |||
XM_005258392.4 | Intron | XP_005258449.1 | |||
XM_006722583.3 | Intron | XP_006722646.1 | |||
XM_006722586.3 | Intron | XP_006722649.1 | |||
XM_006722587.3 | Intron | XP_006722650.1 | |||
XM_006722588.3 | Intron | XP_006722651.1 | |||
XM_006722589.3 | Intron | XP_006722652.1 | |||
XM_011526278.2 | Intron | XP_011524580.1 | |||
XM_011526279.2 | Intron | XP_011524581.1 | |||
XM_017026100.1 | Intron | XP_016881589.1 | |||
XM_017026101.1 | Intron | XP_016881590.1 | |||
XM_017026102.1 | Intron | XP_016881591.1 |