Product Details
- SNP ID
-
rs6003824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23650864 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATTTACCCATTTTAAGTTCACATT[G/T]TAGCGTACAACCATCACCAAAACCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GUSBP11
PubMed Links
Gene Details
- Gene
- GUSBP11
- Gene Name
- glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.
View Full Product Details