Product Details
- SNP ID
-
rs9939246
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:58467758 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAAAATGGGATCGTCGTGGTGCCT[A/G]CCTGGCTGGGATCTTTGGAGATTAG
- Phenotype
-
MIM: 614463
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NDRG4
PubMed Links
Gene Details
- Gene
- NDRG4
- Gene Name
- NDRG family member 4
View Full Product Details