Product Details

SNP ID
rs7528405
Assay Type
Functionally tested
NCBI dbSNP Submissions
74
Location
Chr.1:21981045 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCACCTCCGCCCGCAGGAGCTCC[C/T]GGACCTACCAGGTGGTGTTGGGCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELA3B PubMed Links

Gene Details

Gene
CELA3B
Gene Name
chymotrypsin like elastase family member 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007352.3 381 Missense Mutation CGG,TGG R79W NP_031378.1

View Full Product Details